missing translation for 'onlineSavingsMsg'
Learn More

N-Benzoylthiourea, 98%

Product Code. 11368775
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11368775 5 g 5g
11378775 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11368775 Supplier Thermo Scientific Alfa Aesar Supplier No. L12377.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 614-23-3 | C8H8N2OS | 180.225 g/mol

N-Benzoylthiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzoylthiourea is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 614-23-3
Molecular Formula C8H8N2OS
Molecular Weight (g/mol) 180.225
MDL Number MFCD00041191
InChI Key DQMWMUMCNOJLSI-UHFFFAOYSA-N
Synonym benzoylthiourea, n-benzoylthiourea, 1-benzoyl-2-thiourea, necacyl, 1-benzoylthiourea, benzamide, n-thiocarbamoyl, urea, 1-benzoyl-2-thio, benzamide, n-aminothioxomethyl, n-aminothioxomethyl benzamide, benzamide, n-aminothioxomethyl-9ci
PubChem CID 2735473
IUPAC Name N-carbamothioylbenzamide
SMILES C1=CC=C(C=C1)C(=O)NC(=S)N

Specifications

Melting Point 174°C to 176°C
Odor Odorless
Quantity 5 g
Beilstein 1942772
Solubility Information Insoluble in water.
Formula Weight 180.23
Percent Purity 98%
Chemical Name or Material N-Benzoylthiourea

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.