missing translation for 'onlineSavingsMsg'
Learn More

N-Benzyl-tert-butylamine, 96%

Product Code. p-7043528
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11394474

Brand: Thermo Scientific Alfa Aesar L01146.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3378-72-1 | C11H17N | 163.264 g/mol

N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzyl-tert-butylamine, is used as a protected amine. It aids in the oxidation to the corresponding hydroxylamine and nitrone.

Solubility
Insoluble in water.

Notes
Air Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from oxidizing agents. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3378-72-1
Molecular Formula C11H17N
Molecular Weight (g/mol) 163.264
MDL Number MFCD00008798
InChI Key DLSOILHAKCBARI-UHFFFAOYSA-N
Synonym n-tert-butyl benzylamine, n-tert-butylbenzylamine, n-benzyl-tert-butylamine, tert-butylbenzylamine, n-t-butylbenzylamine, benzenemethanamine, n-1,1-dimethylethyl, benzyl tert-butyl amine, tert-butyl benzylamine, t-butylbenzylamine, n-1,1-dimethylethyl benzylamine
PubChem CID 76908
IUPAC Name N-benzyl-2-methylpropan-2-amine
SMILES CC(C)(C)NCC1=CC=CC=C1

Specifications

Melting Point -30°C to -25°C
Density 0.899
Boiling Point 218°C to 220°C
Flash Point 88°C (190°F)
Linear Formula C6H5CH2NHC(CH3)3
Refractive Index 1.497
Quantity 25 g
UN Number UN2735
Beilstein 507464
Sensitivity Air Sensitive
Solubility Information Insoluble in water.
Formula Weight 163.27
Percent Purity 96%
Chemical Name or Material N-Benzyl-tert-butylamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.