missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylaminoacetaldehyde diethyl acetal, 96%

Product Code. 11471488
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11471488 25 g 25g
11451488 1 g 1g
11461488 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11471488 Supplier Thermo Scientific Alfa Aesar Supplier No. A13841.14
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 61190-10-1 | C13H21NO2 | 223.316 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61190-10-1
Molecular Formula C13H21NO2
Molecular Weight (g/mol) 223.316
MDL Number MFCD00051599
InChI Key SXFVQTYQHWRYOS-UHFFFAOYSA-N
Synonym n-benzylaminoacetaldehyde diethyl acetal, benzyl 2,2-diethoxyethyl amine, n-benzyl-n-2,2-diethoxyethyl amine, 2,2-diethoxyethyl benzylamine, acmc-20ak6r, benzylaminoacetaldehyde diethylacetal, benzyl-2,2-diethoxy-ethyl-amine, benzylamino-acetaldehyde diethyl acetal, n-benzyl 2-aminoacetaldehyde diethyl acetal
PubChem CID 521961
IUPAC Name N-benzyl-2,2-diethoxyethanamine
SMILES CCOC(CNCC1=CC=CC=C1)OCC

Specifications

Boiling Point 115°C to 117°C (1 mmHg)
Refractive Index 1.489
Quantity 25 g
Beilstein 2694610
Formula Weight 223.32
Percent Purity 96%
Chemical Name or Material N-Benzylaminoacetaldehyde diethyl acetal

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.