missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylethanolamine, 96%

Produktkod. 11442898
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2500 g
Förpackningsstorlek:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity Förpackningsstorlek
11442898 500 g 500g
11452898 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 missing translation for 'options'
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11442898 missing translation for 'mfr' Thermo Scientific Alfa Aesar missing translation for 'supplierNo' A16866.36
 Restricted Item
Estimated Shipment: 20-11-2026
Lägg i varukorgen
Lägg i varukorgen
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denna artikel kan inte returneras. Se returpolicy

CAS: 104-63-2 | C9H13NO | 151.21 g/mol

Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

Solubility
Soluble in water 162 g/L (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents..
TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 104-63-2
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002840
InChI Key XNIOWJUQPMKCIJ-UHFFFAOYSA-N
Synonym n-benzylethanolamine, 2-benzylamino ethanol, benzylaminoethanol, benzylethanolamine, ethanol, 2-phenylmethyl amino, 2-benzylamino ethan-1-ol, benzyl ethanolamine, n-benzyle ethanol amine, ethanol, 2-benzylamino, unii-48121ms9jm
PubChem CID 4348
IUPAC Name 2-(benzylamino)ethanol
SMILES OCCNCC1=CC=CC=C1

Specifikationer

Melting Point -30°C
Density 1.063
pH 11
Boiling Point 240°C
Flash Point 130°C (266°F)
Refractive Index 1.543
Quantity 500 g
Beilstein 775164
Solubility Information Soluble in water 162g/L (25°C).
Formula Weight 151.21
Percent Purity 96%
Chemical Name or Material N-Benzylethanolamine
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.