missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylethanolamine, 96%

Product Code. 11452898
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
This item is not returnable. View return policy

Product Code. 11452898

Brand: Thermo Scientific Alfa Aesar A16866.0E

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 104-63-2 | C9H13NO | 151.21 g/mol

Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Has Pharmaceutical applications. N-Benzylethanolamine is used in the synthesis of imidazoles as potent calcitonin (CGRP) antagonists. Also it aids in the preparation of benzofused hydroxamix acids, as useful fragments for the synthesis of histone deacetylase inhibitors.

Solubility
Soluble in water 162 g/L (25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agents..
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 104-63-2
Molecular Formula C9H13NO
Molecular Weight (g/mol) 151.21
MDL Number MFCD00002840
InChI Key XNIOWJUQPMKCIJ-UHFFFAOYSA-N
Synonym n-benzylethanolamine, 2-benzylamino ethanol, benzylaminoethanol, benzylethanolamine, ethanol, 2-phenylmethyl amino, 2-benzylamino ethan-1-ol, benzyl ethanolamine, n-benzyle ethanol amine, ethanol, 2-benzylamino, unii-48121ms9jm
PubChem CID 4348
IUPAC Name 2-(benzylamino)ethanol
SMILES OCCNCC1=CC=CC=C1

Specifications

Melting Point -30°C
Density 1.063
pH 11
Boiling Point 240°C
Flash Point 130°C (266°F)
Refractive Index 1.543
Quantity 2500 g
Beilstein 775164
Solubility Information Soluble in water 162g/L (25°C).
Formula Weight 151.21
Percent Purity 96%
Chemical Name or Material N-Benzylethanolamine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.