missing translation for 'onlineSavingsMsg'
Learn More

N-Benzylmethylamine, 97%

Product Code. 11454784
Change view
Click to view available options
Quantity:
100 mL
250 mL
1000 mL
Unit Size:
1000mL
100mL
250mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454784 250 mL 250mL
11468760 100 mL 100mL
11444784 1000 mL 1000mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11454784 Supplier Thermo Scientific Alfa Aesar Supplier No. A13746.AK
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 103-67-3 | C8H11N | 121.18 g/mol

N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Benzylmethylamine is used in the production of dyes. It can be also used as catalytic agent, Petrochemical additive.

Solubility
Soluble in water (65 g/L at 20°C).

Notes
Air Sensitive. Incompatible with strong oxidizing agents, acids, acid chlorides, acid anhydrides, carbon dioxide.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 103-67-3
Molecular Formula C8H11N
Molecular Weight (g/mol) 121.18
MDL Number MFCD00008289
InChI Key RIWRFSMVIUAEBX-UHFFFAOYSA-N
Synonym n-methylbenzylamine, methylbenzylamine, n-benzylmethylamine, benzyl methyl amine, benzenemethanamine, n-methyl, benzylmethylamine, n-methy-n-benzylamine, n-benzyl-n-methylamine, benzylamine, n-methyl, n-methyl phenyl methanamine
PubChem CID 7669
IUPAC Name N-methyl-1-phenylmethanamine
SMILES CNCC1=CC=CC=C1

Specifications

Melting Point -55°C
Density 0.94
pH 10
Boiling Point 184°C to 188°C
Flash Point 77°C (170°F)
Odor Amine-like
Linear Formula C6H5CH2NHCH3
Refractive Index 1.523
Quantity 250 mL
UN Number UN2922
Beilstein 606221
Sensitivity Air Sensitive
Solubility Information Soluble in water (65g/L at 20°C).
Formula Weight 121.18
Percent Purity 97%
Chemical Name or Material N-Benzylmethylamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.