missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-3,3-diphenyl-L-alanine, 95%

Product Code. 15498567
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498567 5 g 5g
15408577 250 mg 250mg
15488567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15498567 Supplier Thermo Scientific Alfa Aesar Supplier No. H52063.06
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 138662-63-2 | C20H23NO4 | 341.407 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 138662-63-2
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.407
MDL Number MFCD00191186
InChI Key TYJDOLCFYZSNQC-KRWDZBQOSA-N
Synonym boc-l-3,3-diphenylalanine, boc-3,3-diphenyl-l-alanine, s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, s-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-l-phenylalanine, boc-beta-phenyl-phe-oh, n-tert-butoxycarbonyl-beta-phenyl-l-phenylalanine, s-2-tert-butoxycarbonylamino-3,3-diphenylpropanoic acid, 2s-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, pubchem20362
PubChem CID 2761487
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3,3-diphenylpropanoic acid
SMILES CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C2=CC=CC=C2)C(=O)O

Specifications

Melting Point 160°C (decomposition)
Quantity 5 g
Beilstein 4880641
Formula Weight 341.41
Percent Purity 95%
Chemical Name or Material N-Boc-3,3-diphenyl-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.