missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-beta-phenyl-D-phenylalanine, 98%

Product Code. 15493438
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15493438 500 mg 500mg
15483438 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15493438 Supplier Thermo Scientific Alfa Aesar Supplier No. H56885.ME
 Restricted Item
Estimated Shipment: 12-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 143060-31-5 | C20H23NO4 | 341.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 143060-31-5
Molecular Formula C20H23NO4
Molecular Weight (g/mol) 341.41
MDL Number MFCD00191187
InChI Key TYJDOLCFYZSNQC-UHFFFAOYNA-N
Synonym boc-d-3,3-diphenylalanine, boc-d-ala 3,3-diphenyl-oh, r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, boc-3,3-diphenyl-d-alanine, r-n-boc-2-amino-3,3-diphenylpropionic acid, n-boc-beta-phenyl-d-phenylalanine, n-tert-butoxycarbonyl-beta-phenyl-d-phenylalanine, 2r-2-tert-butoxycarbonyl amino-3,3-diphenylpropanoic acid, n-tert-butoxycarbonyl-3-phenyl-l-phenylalanine, n-boc-b-phenyl-d-phenylalanine
PubChem CID 7019136
SMILES CC(C)(C)OC(=O)NC(C(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Melting Point 157°C
Quantity 500 mg
Optical Rotation −30 (c=1 in Ethanol)
IUPAC Name 2-{[(tert-butoxy)carbonyl]amino}-3,3-diphenylpropanoic acid
Formula Weight 341.4
Percent Purity 98%
Chemical Name or Material N-Boc-beta-phenyl-D-phenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.