missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-DL-2-Aminotetralin-2-carboxylic acid, 98%, Thermo Scientific™

Product Code. 10143631 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
10143631 1 g 1g
10026243 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 10143631

Brand: Thermo Scientific Acros 334370010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 98569-12-1
MDL Number MFCD00800586
InChI Key DSDQZWPRZHNCIX-UHFFFAOYSA-N
Synonym 2-boc-amino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 2-n-boc-amino-tetrahydro-2-naphthoic acid, 2-tert-butoxycarbonylamino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, n-boc-dl-2-aminotetralin-2-carboxylic acid, 2-n-boc-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 2-tert-butoxycarbonyl amino-3,4-dihydro-1h-naphthalene-2-carboxylic acid, 2-tert-butoxy carbonylamino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, 2-tert-butoxycarbonyl amino-1,2,3,4-tetrahydronaphthalene-2-carboxylic acid, boc-atc
PubChem CID 2733177
IUPAC Name 2-[(2-methylpropan-2-yl)oxycarbonylamino]-3,4-dihydro-1H-naphthalene-2-carboxylic acid
SMILES CC(C)(C)OC(=O)NC1(CCC2=CC=CC=C2C1)C(=O)O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 177°C to 187°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Molecular Formula C16H21NO4
Quantity 1 g
Solubility Information Solubility in water: soluble. Other solubilities: soluble in methanol,chloroform
Molecular Weight (g/mol) 291.35
Formula Weight 291.35
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-BOC-DL-2-Aminotetralin-2-carboxylic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.