missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-ethylenediamine, 98%, may cont up to 5% tert-butanol

Product Code. 11379955
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11379955 25 g 25g
11369955 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11379955 Supplier Thermo Scientific Alfa Aesar Supplier No. L19947.14
 Restricted Item
Estimated Shipment: 22-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 57260-73-8 | C7H16N2O2 | 160.22 g/mol

N-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-ethylenediamine is used in the synthesis of thyronamine derivatives. It is also used in the preparation of (2-isothiocyanato-ethyl)-carbamic acid tert-butyl ester by reacting with carbon disulfide. Further, it is used in both oligonucleotide and peptide synthesis.

Solubility
Miscible with methanol and chloroform. Slightly miscible with water.

Notes
Air sensitive. Store in a cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 57260-73-8
Molecular Formula C7H16N2O2
Molecular Weight (g/mol) 160.22
MDL Number MFCD00191871
InChI Key AOCSUUGBCMTKJH-UHFFFAOYSA-N
Synonym n-boc-ethylenediamine, tert-butyl n-2-aminoethyl carbamate, tert-butyl 2-aminoethyl carbamate, n-t-boc-ethylenediamine, n-tert-boc-ethylenediamine, tert-butyl 2-aminoethylcarbamate, n-boc-1,2-diaminoethane, n-boc-ethane-1,2-diamine, mono-n-t-boc-eda, n-boc-diaminoethane
PubChem CID 187201
IUPAC Name tert-butyl N-(2-aminoethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCN

Specifications

Boiling Point 72°C (0.1mmHg)
Refractive Index 1.457
UN Number UN2735
Beilstein 1932330
Solubility Information Miscible with methanol and chloroform. Slightly miscible with water.
Density 1.016
Quantity 25 g
Flash Point >110°C (230°F)
Formula Weight 160.22
Sensitivity Air Sensitive
Percent Purity 98%
Assay may cont up to 5% tert-butanol
Chemical Name or Material N-Boc-ethylenediamine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.