missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-N-desmethyl Dapoxetine, TRC

Product Code. 30333633
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30333633 5 mg 5mg
30341510 10 mg 10mg
30340001 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30333633 Supplier Toronto Research Chemicals Supplier No. TRCB6464005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-Boc-N-desmethyl Dapoxetine
Analyte or Component Names N-Boc-N-desmethyl Dapoxetine
Shipping Condition Room Temperature
Molecular Formula C25 H29 N O3
Synonym tert-Butyl Methyl(3-(naphthalen-1-yloxy)-1-phenylpropyl)carbamate
InChI Formula InChI=1S/C25H29NO3/c1-25(2,3)29-24(27)26(4)22(20-12-6-5-7-13-20)17-18-28-23-16-10-14-19-11-8-9-15-21(19)23/h5-16,22H,17-18H2,1-4H3
Solution Type Neat
SMILES CN(C(CCOc1cccc2ccccc12)c3ccccc3)C(=O)OC(C)(C)C
IUPAC Name tert-butyl N-methyl-N-(3-naphthalen-1-yloxy-1-phenylpropyl)carbamate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 391.503
Quantity 5 mg
Formula Weight 391.215
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.