missing translation for 'onlineSavingsMsg'
Learn More

N-α-BOC-N-omega-Tosyl-L-arginine, 97%, contains <15% ethyl acetate, Thermo Scientific™

Product Code. 11975501 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11975501 5 g 5g
11995501 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11975501 Supplier Thermo Scientific Acros Supplier No. 440830050
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 13836-37-8 , 141-78-6
Molecular Formula C18H28N4O6S
Molecular Weight (g/mol) 428.51
InChI Key WBIIPXYJAMICNU-AWEZNQCLSA-N
Synonym boc-arg tos-oh, boc-arg tos, n alpha-boc-n omega-tosyl-l-arginine, n 2-tert-butoxycarbonyl-n g-tosyl-l-arginine, 2s-2-tert-butoxycarbonyl amino-5-n'-4-methylbenzenesulfonyl carbamimidamido pentanoic acid, l-ornithine, n2-1,1-dimethylethoxy carbonyl-n5-imino 4-methylphenyl sulfonyl amino methyl, boc-arg tos oh, boc-arg ts, ambotzbaa1068, boc arg tos-oh
PubChem CID 83767
IUPAC Name (2S)-5-[[amino-[(4-methylphenyl)sulfonylamino]methylidene]amino]-2-[(2-methylpropan-2-yl)oxycarbonylamino]pentanoic acid
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(=NCCCC(C(=O)O)NC(=O)OC(C)(C)C)N

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 99°C to 100°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (HPLC)
Quantity 5 g
Specific Rotation -3.2
Residual Solvents 15.0% max. (EtOAc)
Formula Weight 428.51
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-α-BOC-N-ω-Tosyl-L-arginine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.