missing translation for 'onlineSavingsMsg'
Learn More

N-BOC-O-Benzyl-D-serine, 98%

Product Code. 10343342 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10343342

Brand: Thermo Scientific Acros 334670050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 47173-80-8 | C15H20NO5 | 294.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47173-80-8
Molecular Formula C15H20NO5
Molecular Weight (g/mol) 294.33
MDL Number MFCD00038248
InChI Key DMBKPDOAQVGTST-GFCCVEGCSA-M
Synonym n-boc-o-benzyl-d-serine, boc-d-ser bzl-oh, boc-o-benzyl-d-serine, o-benzyl-n-tert-butoxycarbonyl-d-serine, d-serine, n-1,1-dimethylethoxy carbonyl-o-phenylmethyl, 2r-3-benzyloxy-2-tert-butoxycarbonyl amino propanoic acid, 2r-3-benzyl-oxy-2-tert-butoxy carbonyl amino propanoic acid, pubchem12444, pubchem14944, boc-d-ser obn-oh
PubChem CID 2733693
SMILES CC(C)(C)OC(=O)N[C@H](COCC1=CC=CC=C1)C([O-])=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 58°C to 61°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Quantity 5 g
Specific Rotation Condition −19.00 (20.00°C c=2 80% alcohol)
Specific Rotation -19
IUPAC Name (2R)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-phenylmethoxypropanoic acid
Formula Weight 295.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-BOC-O-Benzyl-D-serine, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.