missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-O-tert-butyl-L-tyrosine, 98%

Product Code. 15489268
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15489268 5 g 5g
15499268 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15489268 Supplier Thermo Scientific Alfa Aesar Supplier No. H62801.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 47375-34-8 | C18H27NO5 | 337.42 g/mol

N-Boc-O-tert-butyl-L-tyrosine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Boc-O-tert-butyl-L-tyrosine is used as pharmaceutical intermediate.

Solubility
Soluble in DMF. Insoluble in water.

Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents, heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 47375-34-8
Molecular Formula C18H27NO5
Molecular Weight (g/mol) 337.42
MDL Number MFCD00065598
InChI Key ZEQLLMOXFVKKCN-YQTOOIBONA-N
Synonym boc-tyr tbu-oh, boc-o-tert-butyl-l-tyrosine, boc-tyr tbul-oh, s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, n-tert-butoxycarbonyl-o-tert-butyl-l-tyrosine, l-tyrosine, n-1,1-dimethylethoxy carbonyl-o-1,1-dimethylethyl, 2s-3-4-tert-butoxy phenyl-2-tert-butoxycarbonyl amino propanoic acid, 2s-2-tert-butoxy carbonylamino-3-4-tert-butoxy phenyl propanoic acid, boc-tyr but-oh, pubchem12288
PubChem CID 7017901
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-[4-[(2-methylpropan-2-yl)oxy]phenyl]propanoic acid
SMILES CC(C)(C)OC(=O)N[C@@H](CC1=CC=C(OC(C)(C)C)C=C1)C(O)=O

Specifications

Melting Point 113°C to 118°C
Quantity 5 g
Beilstein 4327336
Solubility Information Soluble in DMF. Insoluble in water.
Optical Rotation −15° (c=1 in DMF)
Formula Weight 337.41
Percent Purity 98%
Chemical Name or Material N-Boc-O-tert-butyl-L-tyrosine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.