missing translation for 'onlineSavingsMsg'
Learn More

N-Boc-S-trityl-D-cysteine, 98%

Product Code. 15429318
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15429318 1 g 1g
15439318 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15429318 Supplier Thermo Scientific Alfa Aesar Supplier No. H62863.03
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 87494-13-1 | C27H29NO4S | 463.59 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 87494-13-1
Molecular Formula C27H29NO4S
Molecular Weight (g/mol) 463.59
MDL Number MFCD00236839
InChI Key JDTOWOURWBDELG-UHFFFAOYNA-N
Synonym boc-d-cys trt-oh, boc-s-trityl-d-cysteine, n-alpha-t-butyloxycarbonyl-s-trityl-d-cysteine, s-2-tert-butoxycarbonyl amino-3-tritylthio propanoic acid, boc-l-cysteine trityl, 2s-2-tert-butoxycarbonyl amino-3-triphenylmethyl sulfanyl propanoic acid, boc-cys trt, boc-cys-trt, ambotzbaa5000, n-boc-s-trityl-d-cysteine
PubChem CID 11590774
SMILES CC(C)(C)OC(=O)NC(CSC(C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1)C(O)=O

Specifications

Quantity 1 g
IUPAC Name (2S)-2-[(2-methylpropan-2-yl)oxycarbonylamino]-3-tritylsulfanylpropanoic acid
Formula Weight 463.59
Percent Purity 98%
Chemical Name or Material N-Boc-S-trityl-D-cysteine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.