missing translation for 'onlineSavingsMsg'
Learn More

Thimerosal

Product Code. 11458760
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11458760 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11458760 Supplier Thermo Scientific Alfa Aesar Supplier No. A14043.22
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 94-26-8 | C11H14O3 | 194.23 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
It is used as an preservative in cosmetics such as eye shadow, foundation, sunscreen, facial moisturizer and skin anti-aging treatment. It is also used in medication suspensions, and as a flavoring additive in food.

Solubility
Soluble in DMSO, ethyl acetate, methanol.

Notes
Store in refrigerator. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 94-26-8
Melting Point 67°C to 71°C
Molecular Formula C11H14O3
MDL Number MFCD00016478
Quantity 100 g
Beilstein 1103741
Merck Index 14,1584
Synonym butylparaben, butyl paraben, butyl p-hydroxybenzoate, nipabutyl, butoben
Solubility Information Soluble in DMSO,ethyl acetate,methanol.
InChI Key QFOHBWFCKVYLES-UHFFFAOYSA-N
SMILES CCCCOC(=O)C1=CC=C(O)C=C1
IUPAC Name butyl 4-hydroxybenzoate
Molecular Weight (g/mol) 194.23
PubChem CID 7184
Formula Weight 194.23
Percent Purity ≥99%
Chemical Name or Material n-Butyl 4-hydroxybenzoate
Show More Show Less

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.