missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Cadherin / Cadherin-2 / CD325 Mouse, Clone: CDH2/1573, enQuireBio™
Mouse Monoclonal Antibody
199.00 € - 478.00 €
Specifications
| Antigen | N-Cadherin / Cadherin-2 / CD325 |
|---|---|
| Clone | CDH2/1573 |
| Concentration | 200μg/mL |
| Applications | Immunohistochemistry |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
15949697
|
EnquireBio
1000-MSM3-P0 |
20 μg | |||||||
|
15959697
|
EnquireBio
1000-MSM3-P1 |
100 μg |
554.40 € 478.00 € / 100µg Save 76.40 € 13% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
| N-Cadherin / Cadherin-2 / CD325 | |
| 200μg/mL | |
| Monoclonal | |
| Mouse | |
| 10mM PBS with 0.05% BSA and 0.05% Azide | |
| 1000 | |
| Recombinant human CDH2 intracellular domain | |
| Primary | |
| Short term: Store at 2-4°C. Long term, aliquot and store at -20°C. | |
| 130-140kDa |
| CDH2/1573 | |
| Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| NP_001295105 | |
| CDH2 | |
| IgG1 κ | |
| Protein A/G Purified | |
| Cell surface | |
| HeLa or HUVEC cells. Heart, Pancreas or Cerebral Cortex |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title