missing translation for 'onlineSavingsMsg'
Learn More

N-CBZ-D-glutamic acid α-benzyl ester, 98%, Thermo Scientific™

Product Code. 11905521 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11905521

Brand: Thermo Scientific Acros 440940010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 65706-99-2
Molecular Formula C20H21NO6
Molecular Weight (g/mol) 371.39
InChI Key VWHKODOUMSMUAF-QGZVFWFLSA-N
Synonym z-d-glu-obzl, cbz-d-glu-obzl, n-cbz-d-glutamic acid alpha-benzyl ester, r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, 1-benzyl n-carbobenzoxy-d-glutamate, 4r-5-benzyloxy-4-benzyloxy carbonyl amino-5-oxopentanoic acid, z-d-glu-obn, cbz-d-glu-och2ph, pubchem14966, cbz-d-glu oh-obzl
PubChem CID 7018162
IUPAC Name (4R)-5-oxo-5-phenylmethoxy-4-(phenylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C(C=C1)COC(=O)C(CCC(=O)O)NC(=O)OCC2=CC=CC=C2

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 97°C to 98°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Specific Rotation 24
Formula Weight 371.39
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-CBZ-D-glutamic acid α-benzyl ester
Show More Show Less
Product Identifier
  • N-Cbz-D-Glutamic acid alpha-benzyl ester

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.