missing translation for 'onlineSavingsMsg'
Learn More

n-Decane 1000 μg/mL in Methanol, Dr. Ehrenstorfer

Product Code. 30312086
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30312086 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30312086 Supplier Dr. Ehrenstorfer Supplier No. DREGA09010087ME
 Restricted Item
Estimated Shipment: 13-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifica

Chemical Name or Material n-Decane
CAS 124-18-5
Analyte or Component Names n-Decane
Shipping Condition Room Temperature
Molecular Formula C10 H22
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Decane, Cactus Normal Paraffin N 10, NSC 8781, n-Decane
InChI Formula InChI=1S/C10H22/c1-3-5-7-9-10-8-6-4-2/h3-10H2,1-2H3
SMILES CCCCCCCCCC
IUPAC Name decane
Solvent or Matrix Methanol
Molecular Weight (g/mol) 142.28
Quantity 1 mL
Formula Weight 142.1722 g/mol
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.