missing translation for 'onlineSavingsMsg'
Learn More

N-desmethyl Enzalutamide, MedChemExpress

Product Code. 16460618
Encompass_Preferred
missing translation for 'changeView'
missing translation for 'orderingAttributeHoverText'
Quantity:
5 mg
10 mg
50 mg
100 mg
missing translation for 'unitSize':
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity missing translation for 'unitSize'
16460618 5 mg 5mg
16438168 10 mg 10mg
16417926 100 mg 100mg
16404077 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 missing translation for 'options'
This item is not returnable. View return policy
Product Code. 16460618 missing translation for 'mfr' Medchem Express missing translation for 'supplierNo' HY70002A
 Restricted Item
Estimated Shipment: 29-09-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress N-desmethyl Enzalutamide is the active metabolite of Enzalutamide.N-desmethyl Enzalutamide is the active metabolite of Enzalutamide. N-desmethyl Enzalutamide demonstrates primary and secondary pharmacodynamics of similar potency to Enzalutamide and circulates at approximately the same plasma concentrations as enzalutamide.

N-desmethyl Enzalutamide is the active metabolite of Enzalutamide.N-desmethyl Enzalutamide is the active metabolite of Enzalutamide. N-desmethyl Enzalutamide demonstrates primary and secondary pharmacodynamics of similar potency to Enzalutamide and circulates at approximately the same plasma concentrations as enzalutamide.

Specifications

Chemical Name or Material N-desmethyl Enzalutamide
CAS 1242137-16-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H14F4N4O2S
Quantity 5 mg
Synonym N-desmethyl MDV 3100
Solubility Information DMSO : 100 mg/mL (222.02 mM; Need ultrasonic)
SMILES S=C(N1C2=CC(F)=C(C(N)=O)C=C2)N(C3=CC=C(C(C(F)(F)F)=C3)C#N)C(C(C)1C)=O
Molecular Weight (g/mol) 450.41
Formula Weight 450.41
Percent Purity 96.97%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.