missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-p-toluidine, 97%, Thermo Scientific™

Product Code. 15244854 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15244854 25 g 25g
15234854 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15244854 Supplier Thermo Scientific Acros Supplier No. 450780250
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 622-57-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
InChI Key AASABFUMCBTXRL-UHFFFAOYSA-N
Synonym n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
PubChem CID 61164
IUPAC Name N-ethyl-4-methylaniline
SMILES CCNC1=CC=C(C=C1)C

Specifications

Density 0.9380g/mL
Boiling Point 218°C
Flash Point 89°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NHC2H5
Quantity 25 g
Specific Gravity 0.938
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material N-Ethyl-p-toluidine
Show More Show Less
compliance-icons
Product Identifier
  • N-Ethyl-p-toluidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.