missing translation for 'onlineSavingsMsg'
Learn More

N-Ethyl-p-toluidine, 97%, Thermo Scientific™

Product Code. p-4813575 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 15244854

Brand: Thermo Scientific Acros 450780250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 622-57-1
Molecular Formula C9H13N
Molecular Weight (g/mol) 135.21
InChI Key AASABFUMCBTXRL-UHFFFAOYSA-N
Synonym n-ethyl-p-toluidine, benzenamine, n-ethyl-4-methyl, n-ethyl-4-toluidine, n-ethyl-p-methylaniline, p-methyl-n-ethylaniline, p-toluidine, n-ethyl, n-ethyl-4-methyl-aniline, n-athyl-p-toluidin, 4-ethylamino toluene, ethyl 4-methylphenyl amine
PubChem CID 61164
IUPAC Name N-ethyl-4-methylaniline
SMILES CCNC1=CC=C(C=C1)C

Specifications

Density 0.9380g/mL
Boiling Point 218°C
Flash Point 89°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH3C6H4NHC2H5
Quantity 25 g
Specific Gravity 0.938
Formula Weight 135.21
Percent Purity 97%
Chemical Name or Material N-Ethyl-p-toluidine
Show More Show Less
compliance-icons
Product Identifier
  • N-Ethyl-p-toluidine
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H310-Fatal in contact with skin.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H330-Fatal if inhaled.
  • H335-May cause respiratory irritation.
  • H300-Fatal if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.