missing translation for 'onlineSavingsMsg'
Learn More

N-Fluorobenzenesulfonimide, 97%

Product Code. 11308855
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308855 1 g 1g
11318855 5 g 5g
11328855 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11308855 Supplier Thermo Scientific Alfa Aesar Supplier No. L13955.03
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 133745-75-2 | C12H10FNO4S2 | 315.333 g/mol

N-Fluorobenzenesulfonimide fluorinates aromatics, enolates, azaenolates and carbanions in high yield. It is used for diastereoselective fluorination of Li enolates of chiral carboximides, for directed fluorination of ortho-lithiated aromatics and for a review of electrophilic N-F fluorinating agents. Asymmteric fluorinations is effected in the presence of a Pd-BINAP catalyst system at room temperature in an ionic liquid. It is a reagent employed in a palladium-catalyzed enantioselective fluorination of t-butoxycarbonyl lactones and lactams. It is also used in the electrophilic difluorination of dihalopyridines with butyl lithium and in the direct conversion of alcohols to dibenzenesulfonamides with triphenylphosphine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fluorobenzenesulfonimide fluorinates aromatics, enolates, azaenolates and carbanions in high yield. It is used for diastereoselective fluorination of Li enolates of chiral carboximides, for directed fluorination of ortho-lithiated aromatics and for a review of electrophilic N-F fluorinating agents. Asymmteric fluorinations is effected in the presence of a Pd-BINAP catalyst system at room temperature in an ionic liquid. It is a reagent employed in a palladium-catalyzed enantioselective fluorination of t-butoxycarbonyl lactones and lactams. It is also used in the electrophilic difluorination of dihalopyridines with butyl lithium and in the direct conversion of alcohols to dibenzenesulfonamides with triphenylphosphine.

Solubility
Very soluble in acetonitrile, dichloromethane or THF and less soluble in toluene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133745-75-2
Molecular Formula C12H10FNO4S2
Molecular Weight (g/mol) 315.333
MDL Number MFCD00144885
InChI Key RLKHFSNWQCZBDC-UHFFFAOYSA-N
Synonym n-fluorobenzenesulfonimide, nfsi, n-fluoro-n-phenylsulfonyl benzenesulfonamide, n-fluorodibenzenesulfonimide, n-fluorobenzensulfonimide, n-fluorobenzenesulphonimide, accufluor nfsi, n-fluorodi benzenesulfonyl amine, bis phenylsulfonyl fluoroamine, n-fluorobis phenylsulfonyl amine
PubChem CID 588007
IUPAC Name N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
SMILES C1=CC=C(C=C1)S(=O)(=O)N(F)S(=O)(=O)C2=CC=CC=C2

Specifications

Melting Point 114°C to 118°C
Quantity 1 g
Beilstein 5348902
Solubility Information Very soluble in acetonitrile,dichloromethane or THF and less soluble in toluene.
Formula Weight 315.34
Percent Purity 97%
Chemical Name or Material N-Fluorobenzenesulfonimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.