missing translation for 'onlineSavingsMsg'
Learn More

N-Fluorobenzenesulfonimide, TRC

Product Code. 30404821
Change view
Click to view available options
Quantity:
500 mg
1 g
2.5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404821 1 g -
30393355 500 mg -
30403741 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30404821 Supplier Toronto Research Chemicals Supplier No. TRCF5968331G
 Restricted Item
Estimated Shipment: 02-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133745-75-2
Molecular Formula C12H10FNO4S2
Molecular Weight (g/mol) 315.34
IUPAC Name N-(benzenesulfonyl)-N-fluorobenzenesulfonamide
SMILES FN(S(=O)(=O)c1ccccc1)S(=O)(=O)c2ccccc2

Specifications

Quantity 1 g
InChI Formula InChI=1S/C12H10FNO4S2/c13-14(19(15,16)11-7-3-1-4-8-11)20(17,18)12-9-5-2-6-10-12/h1-10H
Formula Weight 315.0035
Chemical Name or Material N-Fluorobenzenesulfonimide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.