missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-1-trityl-D-histidine, 98%

Product Code. 15459028
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15459028 25 g 25g
15449028 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15459028 Supplier Thermo Scientific Alfa Aesar Supplier No. H62504.14
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135610-90-1 | C40H33N3O4 | 619.721 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135610-90-1
Molecular Formula C40H33N3O4
Molecular Weight (g/mol) 619.721
MDL Number MFCD00077061
InChI Key XXMYDXUIZKNHDT-DIPNUNPCSA-N
Synonym fmoc-d-his trt-oh, n-fmoc-n'-trityl-d-histidine, d-histidine, n-9h-fluoren-9-ylmethoxy carbonyl-1-triphenylmethyl, 2r-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-1-triphenylmethyl imidazol-4-yl propanoic acid, pubchem18930, fmoc-d-his 1-trt-oh, fmoc-d-his-trt-oh, n-fmoc-1-trityl-d-histidine, fmoc-n-im-trityl-d-histidine, nalpha-fmoc-nim-trityl-d-histidine
PubChem CID 44828577
IUPAC Name (2R)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-(1-tritylimidazol-4-yl)propanoic acid
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)(C3=CC=CC=C3)N4C=C(N=C4)CC(C(=O)O)NC(=O)OCC5C6=CC=CC=C6C7=CC=CC=C57

Specifications

Quantity 25 g
Optical Rotation −80° (c=1 in Chloroform)
Formula Weight 619.71
Percent Purity 98%
Chemical Name or Material N-Fmoc-1-trityl-D-histidine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.