missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(1-naphthyl)-L-alanine, 98%

Product Code. 15468427
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15468427 5 g 5g
15458427 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15468427 Supplier Thermo Scientific Alfa Aesar Supplier No. H51967.06
 Restricted Item
Estimated Shipment: 22-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 96402-49-2 | C28H23NO4 | 437.50 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 96402-49-2
Molecular Formula C28H23NO4
Molecular Weight (g/mol) 437.50
MDL Number MFCD00151914
InChI Key ORWNVJDLEMVDLV-SANMLTNESA-N
Synonym fmoc-3-1-naphthyl-l-alanine, fmoc-1-nal-oh, fmoc-l-1-naphthylalanine, fmoc-beta-1-naphthyl-l-alanine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, fmoc-3-1 naphthyl-l-alanine, fmoc-l-3-1-naphthyl-alanine, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-1-yl propanoic acid, n-fmoc-3-1-naphthyl-l-alanine
PubChem CID 2734474
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-1-ylpropanoic acid
SMILES OC(=O)[C@H](CC1=CC=CC2=CC=CC=C12)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 171°C to 183°C
Quantity 5 g
Beilstein 4829118
Formula Weight 437.49
Percent Purity 98%
Chemical Name or Material N-Fmoc-3-(1-naphthyl)-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.