missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(2-naphthyl)-L-alanine, 95%

Product Code. 15438737
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15438737 250 mg 250mg
15418737 1 g 1g
15428737 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15438737 Supplier Thermo Scientific Alfa Aesar Supplier No. H52179.MD
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 112883-43-9 | C28H22NO4 | 436.49 g/mol

N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-3-(2-naphthyl)-L-alanine is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112883-43-9
Molecular Formula C28H22NO4
Molecular Weight (g/mol) 436.49
MDL Number MFCD00144886
InChI Key JYUTZJVERLGMQZ-SANMLTNESA-M
Synonym fmoc-2-nal-oh, fmoc-3-2-naphthyl-l-alanine, fmoc-l-3-2-naphthyl-alanine, n-fmoc-3-2-naphthyl-l-alanine, fmoc-beta-2-naphthyl-l-alanine, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, 2s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-naphthalen-2-yl propanoic acid, s-n-fmoc-3-2-naphthyl alanine, ambotzfaa1347
PubChem CID 2734452
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-naphthalen-2-ylpropanoic acid
SMILES [O-]C(=O)[C@H](CC1=CC=C2C=CC=CC2=C1)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 250 mg
Beilstein 7668973
Solubility Information Slightly soluble in water.
Formula Weight 437.49
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(2-naphthyl)-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.