missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-3-(3-pyridyl)-L-alanine, 95%

Product Code. 15428557
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15428557 5 g 5g
15438557 250 mg 250mg
15418557 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15428557 Supplier Thermo Scientific Alfa Aesar Supplier No. H52056.06
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 175453-07-3 | C23H20N2O4 | 388.423 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 175453-07-3
Molecular Formula C23H20N2O4
Molecular Weight (g/mol) 388.423
MDL Number MFCD00144887
InChI Key JQLPMTXRCLXOJO-NRFANRHFSA-N
Synonym fmoc-3-3-pyridyl-l-alanine, fmoc-l-3-pyridylalanine, fmoc-3-pal-oh, fmoc-beta-3-pyridyl-ala-oh, fmoc-l-3-3-pyridyl-alanine, fmoc-3-pyridyl-ala-oh, fmoc-3'-pyridyl-l-ala, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-pyridin-3-yl propanoic acid, n-fmoc-3-3-pyridyl-l-alanine, fmoc-d-3-3-pyridyl-alanine
PubChem CID 6957974
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-pyridin-3-ylpropanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CC4=CN=CC=C4)C(=O)O

Specifications

Quantity 5 g
Beilstein 9154190
Formula Weight 388.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-3-(3-pyridyl)-L-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.