missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-beta-alanine, 99%

Product Code. 15446328
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15446328 25 g 25g
15436328 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15446328 Supplier Thermo Scientific Alfa Aesar Supplier No. H59887.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 35737-10-1 | C18H16NO4 | 310.33 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuffs.

Solubility
Soluble in water and 1% acetic acid.

Notes
Desiccate at -20°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35737-10-1
Molecular Formula C18H16NO4
Molecular Weight (g/mol) 310.33
MDL Number MFCD00063328
InChI Key LINBWYYLPWJQHE-UHFFFAOYSA-M
Synonym fmoc-beta-alanine, fmoc-beta-ala-oh, fmoc-b-ala-oh, fmoc-?-ala-oh, n-fmoc-beta-alanine, fmoc-b-alanine, 3-9h-fluoren-9-ylmethoxy carbonyl amino propanoic acid, fmoc-, a-ala-oh, 3-9h-fluoren-9-yl methoxy carbonyl amino propanoic acid, n-9h-fluoren-9-ylmethoxy carbonyl-beta-alanine
PubChem CID 2724630
IUPAC Name 3-(9H-fluoren-9-ylmethoxycarbonylamino)propanoic acid
SMILES [O-]C(=O)CCNC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 142°C to 147°C
Linear Formula FmocNHCH2CH2CO2H
Quantity 25 g
Beilstein 2302327
Solubility Information Soluble in water and 1% acetic acid.
Formula Weight 311.34
Percent Purity 99%
Chemical Name or Material N-Fmoc-beta-alanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.