missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-D-proline, 98+%, may cont. up to ca 5% water

Product Code. 11482060
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11482060

Brand: Thermo Scientific Alfa Aesar H28500.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 101555-62-8 | C20H19NO4 | 337.375 g/mol

In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
In organic synthesis, N-Fmoc-D-proline are often employed as important intermediates in various areas such as peptide synthesis, asymmetric synthesis, medicinal chemistry, and polymer chemistry.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101555-62-8
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.375
MDL Number MFCD00077067
InChI Key ZPGDWQNBZYOZTI-GOSISDBHSA-N
Synonym fmoc-d-pro-oh, fmoc-d-proline, n-9h-fluoren-9-ylmethoxy carbonyl-d-proline, n-fmoc-d-proline, fmoc-d-pro, 2r-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidine-2-carboxylic acid, 2r-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, fmoc-3-pro-oh, ksc908s9r, n-alpha-fmoc-d-proline
PubChem CID 688136
IUPAC Name (2R)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carboxylic acid
SMILES C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)C(=O)O

Specifications

Melting Point 110°C to 116°C
Quantity 5 g
Formula Weight 337.38
Percent Purity ≥98%
Assay may contain up to ≈5% water
Chemical Name or Material N-Fmoc-D-proline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.