missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Description
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.
Solubility
Slightly soluble in water.
Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Specifications
Specifications
| Melting Point | 115°C to 120°C |
| Quantity | 1 g |
| Beilstein | 8019724 |
| Solubility Information | Slightly soluble in water. |
| Formula Weight | 439.51 |
| Percent Purity | 95% |
| Chemical Name or Material | N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction