missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester, 95%

Product Code. 15439645
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15439645 250 mg 250mg
15429645 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439645 Supplier Thermo Scientific Alfa Aesar Supplier No. H52188.MD
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 203854-49-3 | C25H29NO6 | 439.508 g/mol

N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester is used as organic chemical synthesis intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 203854-49-3
Molecular Formula C25H29NO6
Molecular Weight (g/mol) 439.508
MDL Number MFCD01863053
InChI Key XPCDWOCHPTYDPV-INIZCTEOSA-N
Synonym fmoc-beta-hoglu otbu-oh, fmoc-l-beta-homoglutamic acid 6-tert-butyl ester, fmoc-l-beta-homoglutamic acid otbu, fmoc-beta-homoglu otbu-oh, s-3-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxy-6-oxohexanoic acid, 3s-6-tert-butoxy-3-9h-fluoren-9-ylmethoxy carbonyl amino-6-oxohexanoic acid, n-beta-9-fluorenylmethoxycarbonyl-l-beta-homoglutamic acid delta-t-butyl ester, ambotzfaa6670, fmoc-b-hoglu otbu-oh, fmoc-l-?-homoglutamic acid otbu
PubChem CID 2761518
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxy]-6-oxohexanoic acid
SMILES CC(C)(C)OC(=O)CCC(CC(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

Melting Point 115°C to 120°C
Quantity 250 mg
Beilstein 8019724
Solubility Information Slightly soluble in water.
Formula Weight 439.51
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homoglutamic acid 6-tert-butyl ester

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.