missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homophenylalanine, 95%, Thermo Scientific Chemicals

Product Code. 15418547
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15418547 1 g 1g
15428547 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15418547 Supplier Thermo Scientific Alfa Aesar Supplier No. H52052.03
 Restricted Item
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 193954-28-8 | Molecular Formula: C25H23NO4 | Molecular Weight: 401.462 g/mol

N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homophenylalanine is used for producing high quality peptides.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193954-28-8
Molecular Formula C25H23NO4
Molecular Weight (g/mol) 401.462
MDL Number MFCD01863055
InChI Key DQNUGHJJKNFCND-SFHVURJKSA-N
Synonym fmoc-l-beta-homophenylalanine, fmoc-b-hophe-oh, fmoc-beta-hophe-oh, fmoc-beta-homophe-oh, 3s-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-phenylbutanoic acid, ambotzfaa1657, fmoc-, a-hophe-oh, 3s-3-9h-fluoren-9-ylmethoxycarbonylamino-4-phenylbutanoic acid, n-fmoc-l-beta-homophenylalanine, fmoc-s-3-amino-4-phenylbutyric acid
PubChem CID 2761539
IUPAC Name (3S)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-phenylbutanoic acid
SMILES C1=CC=C(C=C1)CC(CC(=O)O)NC(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24

Specifications

Quantity 1 g
Beilstein 7726557
Solubility Information Slightly soluble in water.
Formula Weight 401.46
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homophenylalanine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.