missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homoproline, 95%

Product Code. 15448567
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15448567 250 mg 250mg
15438567 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448567 Supplier Thermo Scientific Alfa Aesar Supplier No. H52060.MD
 Restricted Item
Estimated Shipment: 25-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 193693-60-6 | C21H21NO4 | 351.402 g/mol

It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in peptide synthesis and drug screening .

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important organic intermediate used in agrochemicals, pharmaceuticals and dyestuff fields. It is also used in peptide synthesis and drug screening .

Notes
Store away from heat and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 193693-60-6
Molecular Formula C21H21NO4
Molecular Weight (g/mol) 351.402
MDL Number MFCD01863058
InChI Key ZNIGOUDZWCDFFC-AWEZNQCLSA-N
Synonym fmoc-l-beta-homoproline, fmoc-beta-hopro-oh, s-2-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidin-2-yl acetic acid, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidin-2-yl acetic acid, 2-2s-1-9h-fluoren-9-ylmethoxycarbonyl pyrrolidin-2-yl acetic acid, fmoc-d-beta-homoproline, fmoc-hopro-oh, fmoc-beta-homopro-oh, fmoc-l-beta-hpro-oh, fmoc-i 2-hopro-oh
PubChem CID 2761542
IUPAC Name 2-[(2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidin-2-yl]acetic acid
SMILES C1CC(N(C1)C(=O)OCC2C3=CC=CC=C3C4=CC=CC=C24)CC(=O)O

Specifications

Quantity 250 mg
Beilstein 7798553
Formula Weight 351.4
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homoproline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.