missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-beta-homovaline, 95%

Product Code. 15498737
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15498737 1 g 1g
15408747 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15498737 Supplier Thermo Scientific Alfa Aesar Supplier No. H52182.03
 Restricted Item
Estimated Shipment: 30-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 172695-33-9 | C21H23NO4 | 353.42 g/mol

N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Fmoc-L-beta-homovaline is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 172695-33-9
Molecular Formula C21H23NO4
Molecular Weight (g/mol) 353.42
MDL Number MFCD01862853
InChI Key KFDMTAMWOIQTOB-LJQANCHMSA-N
Synonym fmoc-l-beta-homovaline, r-3-9h-fluoren-9-yl methoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-hoval-oh, r-n-fmoc-3-amino-4-methylpentanoic acid, r-3-fmoc-amino-4-methylpentanoic acid, 3r-3-9h-fluoren-9-ylmethoxy carbonyl amino-4-methylpentanoic acid, fmoc-beta-leu-oh, fmoc-l-, a-leucine, fmoc-l-beta-leucine, fmoc-, a-hoval-oh
PubChem CID 2761560
IUPAC Name (3R)-3-(9H-fluoren-9-ylmethoxycarbonylamino)-4-methylpentanoic acid
SMILES CC(C)[C@@H](CC(O)=O)NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Quantity 1 g
Beilstein 7388393
Solubility Information Slightly soluble in water.
Formula Weight 353.42
Percent Purity 95%
Chemical Name or Material N-Fmoc-L-beta-homovaline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.