missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-citrulline, 98%

Product Code. 15489988
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15489988 25 g 25g
15469988 1 g 1g
15479988 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15489988 Supplier Thermo Scientific Alfa Aesar Supplier No. H63342.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 133174-15-9 | C21H23N3O5 | 397.431 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 133174-15-9
Molecular Formula C21H23N3O5
Molecular Weight (g/mol) 397.431
MDL Number MFCD00151943
InChI Key NBMSMZSRTIOFOK-SFHVURJKSA-N
Synonym fmoc-cit-oh, fmoc-l-citrulline, nalpha-fmoc-l-citrulline, s-2-9h-fluoren-9-yl methoxy carbonyl amino-5-ureidopentanoic acid, l-ornithine, n5-aminocarbonyl-n2-9h-fluoren-9-ylmethoxy carbonyl, nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-citrulline, 2s-2-9h-fluoren-9-ylmethoxycarbonylamino-5-ureido-pentanoic acid, 2s-5-carbamoylamino-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, fmoc-citoh, pubchem19002
PubChem CID 2756127
IUPAC Name (2S)-5-(carbamoylamino)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
SMILES C1=CC=C2C(=C1)C(C3=CC=CC=C32)COC(=O)NC(CCCNC(=O)N)C(=O)O

Specifications

Quantity 25 g
Beilstein 7780987
Formula Weight 397.42
Percent Purity 98%
Physical Form Solid
Chemical Name or Material N-Fmoc-L-citrulline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.