missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-norvaline, 98%

Product Code. 15452748
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 15452748

Brand: Thermo Scientific Alfa Aesar H56290.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 135112-28-6 | C20H21NO4 | 339.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135112-28-6
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00155631
InChI Key JBIJSEUVWWLFGV-SFHVURJKSA-N
Synonym fmoc-nva-oh, fmoc-l-norvaline, n-9-fluorenylmethyloxycarbonyl-l-norvaline, n-fmoc-l-norvaline, 2s-2-9h-fluoren-9-ylmethoxy carbonyl amino pentanoic acid, l-norvaline, n-9h-fluoren-9-ylmethoxy carbonyl, fmoc-norvaline, fmoc-l-nva-oh, ambotzfaa1415, pubchem14961
PubChem CID 7016885
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)pentanoic acid
SMILES CCC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C(O)=O

Specifications

Melting Point 151°C to 155°C
Quantity 1 g
Beilstein 5883879
Optical Rotation −21° (c=2 in DMF)
Formula Weight 339.39
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-norvaline

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.