missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-proline, 98%

Product Code. 11464537
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464537 25 g 25g
11454537 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464537 Supplier Thermo Scientific Alfa Aesar Supplier No. B21081.14
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 71989-31-6 | C20H19NO4 | 337.38 g/mol

It is used as pharmaceutical intermediate. Fmoc method is used for the synthesis of the new polymeric support for the preparation of C-terminal peptide amides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as pharmaceutical intermediate. Fmoc method is used for the synthesis of the new polymeric support for the preparation of C-terminal peptide amides.

Solubility
Soluble in methanol. (almost transparency.)

Notes
Store at 2-8°C. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-31-6
Molecular Formula C20H19NO4
Molecular Weight (g/mol) 337.38
MDL Number MFCD00037122
InChI Key ZPGDWQNBZYOZTI-UHFFFAOYNA-N
Synonym fmoc-pro-oh, fmoc-l-proline, fmoc-l-pro-oh, n-fmoc-l-proline, n-9-fluorenylmethoxycarbonyl-l-proline, s-1-9h-fluoren-9-yl methoxy carbonyl pyrrolidine-2-carboxylic acid, n-9h-fluoren-9-ylmethoxy carbonyl-l-proline, fmoc-pro, 2s-1-9h-fluoren-9-ylmethoxy carbonyl pyrrolidine-2-carboxylic acid, 9-fluorenylmethoxycarbonyl-l-proline
PubChem CID 688135
IUPAC Name (2S)-1-(9H-fluoren-9-ylmethoxycarbonyl)pyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCCN1C(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12

Specifications

Melting Point 113°C to 117°C
Quantity 25 g
Beilstein 3596735
Solubility Information Soluble in methanol. (almost transparency.)
Optical Rotation −40° (c=1 in Ethyl Acetate)
Formula Weight 337.38
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-proline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.