missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-L-valine, 98%

Product Code. 11450117
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11450117 5 g 5g
11460117 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11450117 Supplier Thermo Scientific Alfa Aesar Supplier No. B21030.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 68858-20-8 | C20H21NO4 | 339.39 g/mol

It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Fmoc-valine (in addition to the other amino acids) is commonly used to synthesize 4-thiazolidinones (e.g. (E)-5-(4-Ethylbenzylidene)-2-thioxothiazolidin-4-one [E925745]) and 4-metathiazanones as well.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is potentially useful for proteomics studies and solid phase peptide synthesis techniques. Fmoc-valine (in addition to the other amino acids) is commonly used to synthesize 4-thiazolidinones (e.g. (E)-5-(4-Ethylbenzylidene)-2-thioxothiazolidin-4-one [E925745]) and 4-metathiazanones as well.

Solubility
Solubility in methanol gives very faint turbidity.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68858-20-8
Molecular Formula C20H21NO4
Molecular Weight (g/mol) 339.39
MDL Number MFCD00037124
InChI Key UGNIYGNGCNXHTR-SFHVURJKSA-N
Synonym fmoc-l-valine, fmoc-val-oh, fmoc-l-val-oh, n-fmoc-l-valine, n-9h-fluoren-9-ylmethoxy carbonyl-l-valine, n-9-fluorenylmethoxycarbonyl-l-valine, fmoc-valine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-3-methylbutanoic acid, n-alpha-fmoc-l-valine, s-n-9h-fluoren-9-ylmethoxy carbonyl valine
PubChem CID 688217
SMILES CC(C)[C@H](NC(=O)OCC1C2=C(C=CC=C2)C2=C1C=CC=C2)C(O)=O

Specifications

Melting Point 142°C to 147°C
Quantity 5 g
Beilstein 2177443
Optical Rotation −17° (c=1 in DMF)
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-methylbutanoic acid
Formula Weight 339.39
Percent Purity 98%
Chemical Name or Material N-Fmoc-L-valine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.