missing translation for 'onlineSavingsMsg'
Learn More

N-alpha-FMOC-N-epsilon-BOC-L-lysine, 98%

Product Code. 10297450 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10297450 1 g Glass bottle 1g
11915471 10 g Glass bottle 10g
11925471 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10297450 Supplier Thermo Scientific Acros Supplier No. 265220010
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 71989-26-9 | C26H32N2O6 | 468.55 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-26-9
Molecular Formula C26H32N2O6
Molecular Weight (g/mol) 468.54
MDL Number MFCD000371
InChI Key UMRUUWFGLGNQLI-QFIPXVFZSA-N
Synonym fmoc-lys boc-oh, fmoc-lys boc, na-fmoc-ne-boc-l-lysine, n-alpha-fmoc-nepsilon-boc-l-lysine, nepsilon-boc-nalpha-fmoc-l-lysine, nepsilon-tert-butoxycarbonyl-nalpha-9h-fluoren-9-ylmethoxy carbonyl-l-lysine, n-fmoc-n'-boc-l-lysine, n-alpha-fmoc-n-epsilon-tert-boc-l-lysine, n, a-fmoc-n, a-boc-l-lysine, s-2-9h-fluoren-9-yl methoxy carbonyl amino-6-tert-butoxycarbonyl amino hexanoic acid
PubChem CID 2724628
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-6-[(2-methylpropan-2-yl)oxycarbonylamino]hexanoic acid
SMILES CC(C)(C)OC(=O)NCCCCC(C(=O)O)NC(=O)OCC1C2=CC=CC=C2C3=CC=CC=C13

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 129.0°C to 135.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (HPLC)
Packaging Glass bottle
Linear Formula FMOC-NHCH(COOH)(CH2)4NHC(O)OC(CH3)3
Quantity 1 g
Specific Rotation Condition −12° (24°C c=2,DMF)
Specific Rotation − 12.00
Formula Weight 468.54
Percent Purity 98%
Physical Form Powder
Chemical Name or Material N-α-FMOC-N-ε-BOC-L-lysine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.