missing translation for 'onlineSavingsMsg'
Learn More

N-Fmoc-O-tert-butyl-L-serine, 97%

Product Code. 15475738
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15475738 25 g 25g
15465738 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15475738 Supplier Thermo Scientific Alfa Aesar Supplier No. H59422.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 71989-33-8 | C22H24NO5 | 382.44 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71989-33-8
Molecular Formula C22H24NO5
Molecular Weight (g/mol) 382.44
MDL Number MFCD00037127
InChI Key REITVGIIZHFVGU-IBGZPJMESA-M
Synonym fmoc-ser tbu-oh, fmoc-o-tert-butyl-l-serine, fmoc-ser t-bu-oh, fmoc-ser but, fmoc-o-tert.butyl-l-serine, fmoc-ser tbu, fmoc-ser but-oh, pubchem10035, fmoc-l-ser otbu oh, fmoc-l-ser tbu-oh
PubChem CID 2724633
IUPAC Name (2S)-2-(9H-fluoren-9-ylmethoxycarbonylamino)-3-[(2-methylpropan-2-yl)oxy]propanoic acid
SMILES CC(C)(C)OC[C@H](NC(=O)OCC1C2=CC=CC=C2C2=CC=CC=C12)C([O-])=O

Specifications

Melting Point 127°C to 130°C
Quantity 25 g
Beilstein 3632013
Optical Rotation +26° (c=1 in Ethyl Acetate)
Formula Weight 383.44
Percent Purity 98%
Chemical Name or Material N-Fmoc-O-tert-butyl-L-serine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.