missing translation for 'onlineSavingsMsg'
Learn More

N-Isopropyl-4-methylbenzenesulfonamide, 97%

Product Code. 15422288
Change view
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15422288 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15422288 Supplier Thermo Scientific Alfa Aesar Supplier No. H55893.MB
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 21230-07-9 | C10H15NO2S | 213.295 g/mol

N-Isopropyl-4-methylbenzenesulfonamide, N-alkylation of sulfonamides using inactive ethers is used as alkylating reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-Isopropyl-4-methylbenzenesulfonamide, N-alkylation of sulfonamides using inactive ethers is used as alkylating reagents.

Solubility
Soluble in water, ethanol.

Notes
Keep container tightly closed i
TRUSTED_SUSTAINABILITY

Specifications

CAS 21230-07-9
Molecular Formula C10H15NO2S
MDL Number MFCD00543042
Quantity 25 mg
Synonym n-isopropyl-4-methylbenzenesulfonamide, 4-methyl-n-propan-2-yl benzene-1-sulfonamide, n-isopropyl-p-toluenesulfonamide, 4-methyl-n-propan-2-yl benzenesulfonamide, methylethyl 4-methylphenyl sulfonyl amine
Solubility Information Soluble in water,ethanol.
InChI Key NXUHAAIJVABUMJ-UHFFFAOYSA-N
SMILES CC1=CC=C(C=C1)S(=O)(=O)NC(C)C
IUPAC Name 4-methyl-N-propan-2-ylbenzenesulfonamide
Molecular Weight (g/mol) 213.295
PubChem CID 4329528
Formula Weight 213.29
Percent Purity 97%
Chemical Name or Material N-Isopropyl-4-methylbenzenesulfonamide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.