missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methylamine, 95%, Thermo Scientific™

Product Code. 10622875
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10622875 1 g Amber Glass Bottle 1g
10451704 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10622875 Supplier Thermo Scientific Maybridge Supplier No. CC49646DA
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 879896-56-7
Molecular Formula C11H13N3O
Molecular Weight (g/mol) 203.25
MDL Number MFCD09702360
InChI Key LJDBGVKOXDSIQQ-UHFFFAOYSA-N
Synonym n-methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methylamine, benzenemethanamine,n-methyl-2-3-methyl-1,2,4-oxadiazol-5-yl, methyl 2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methyl amine, n-methyl-1-2-3-methyl-1,2,4-oxadiazol-5-yl phenyl methanamine, methyl 2-3-methyl 1,2,4-oxadiazol-5-yl phenyl methyl amine
PubChem CID 24229504
SMILES CNCC1=CC=CC=C1C1=NC(C)=NO1

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
IUPAC Name methyl({[2-(3-methyl-1,2,4-oxadiazol-5-yl)phenyl]methyl})amine
Formula Weight 203.24
Percent Purity 95%
Chemical Name or Material N-methyl[2-(3-methyl-1, 2, 4-oxadiazol-5-yl)phenyl]methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.