missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl(2-methyl-3-furyl)methylamine, 95%, Thermo Scientific™

Product Code. 10366274
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10366274 250 mg Amber Glass Bottle 250mg
10235934 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10366274 Supplier Thermo Scientific Maybridge Supplier No. CC55446CB
 Restricted Item
Estimated Shipment: 04-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 916766-85-3
Molecular Formula C7H11NO
Molecular Weight (g/mol) 125.171
MDL Number MFCD09702377
InChI Key IDODRBWAEHXURE-UHFFFAOYSA-N
Synonym n-methyl 2-methyl-3-furyl methylamine, methyl 2-methylfuran-3-yl methyl amine, n-methyl-2-methylfur-3-yl methylamine, 2-methyl-3-methylamino methyl furan, 3-furanmethanamine,n,2-dimethyl, 3-furanmethanamine, n,2-dimethyl, methyl 2-methyl 3-furyl methyl amine, n-methyl-1-2-methylfuran-3-yl methanamine
PubChem CID 24229548
IUPAC Name N-methyl-1-(2-methylfuran-3-yl)methanamine
SMILES CC1=C(C=CO1)CNC

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 125.17
Percent Purity 95%
Chemical Name or Material N-methyl(2-methyl-3-furyl)methylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.