missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-(1,3-thiazol-2-yl)benzylamine, 95%, Thermo Scientific™

Product Code. 10123964
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10123964 1 g Amber Glass Bottle 1g
10577874 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10123964 Supplier Thermo Scientific Maybridge Supplier No. CC36546DA
 Restricted Item
Estimated Shipment: 17-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 892501-89-2
Molecular Formula C11H12N2S
Molecular Weight (g/mol) 204.291
MDL Number MFCD09025830
InChI Key MNTPWZTYKCKBQX-UHFFFAOYSA-N
Synonym n-methyl-3-1,3-thiazol-2-yl benzylamine, n-methyl-1-3-thiazol-2-yl phenyl methanamine, methyl 3-1,3-thiazol-2-yl phenyl methyl amine, benzenemethanamine,n-methyl-3-2-thiazolyl, n-methyl-1-3-1,3-thiazol-2-yl phenyl methanamine
PubChem CID 18525729
IUPAC Name N-methyl-1-[3-(1,3-thiazol-2-yl)phenyl]methanamine
SMILES CNCC1=CC=CC(=C1)C2=NC=CS2

Specifications

CAS Min % 94.5
CAS Max % 100.0
Assay Percent Range 95%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 204.3
Percent Purity 95%
Chemical Name or Material N-Methyl-3-(1, 3-thiazol-2-yl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.