missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-(1H-pyrazol-1-ylmethyl)benzylamine, 90%, Thermo Scientific™

Product Code. 10082254
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10082254 250 mg Amber Glass Bottle 250mg
10396424 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10082254 Supplier Thermo Scientific Maybridge Supplier No. CC56146CB
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-10-2
Molecular Formula C12H15N3
Molecular Weight (g/mol) 201.273
MDL Number MFCD08690303
InChI Key RXKBCAGRFVIQTK-UHFFFAOYSA-N
Synonym n-methyl-3-1h-pyrazol-1-ylmethyl benzylamine, methyl 3-pyrazol-1-ylmethyl phenyl methyl amine, methyl 3-pyrazolylmethyl phenyl methyl amine, benzenemethanamine,n-methyl-3-1h-pyrazol-1-ylmethyl, 1-3-1h-pyrazol-1-yl methyl phenyl-n-methylmethanamine, n-methyl-1-3-1h-pyrazol-1-yl methyl phenyl methanamine
PubChem CID 18525881
IUPAC Name N-methyl-1-[3-(pyrazol-1-ylmethyl)phenyl]methanamine
SMILES CNCC1=CC=CC(=C1)CN2C=CC=N2

Specifications

CAS Min % 89.5
CAS Max % 100.0
Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.27
Percent Purity 90%
Chemical Name or Material N-methyl-3-(1H-pyrazol-1-ylmethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.