missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-3-phenylpropylamine, TRC

Product Code. 30009989
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009989 25 mg 25mg
30010406 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30009989 Supplier Toronto Research Chemicals Supplier No. TRCM32569025MG
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23580-89-4
Molecular Formula C10 H15 N
Molecular Weight (g/mol) 149.23
Synonym N-methyl-3-phenyl-propan-1-amine, Fluoxetine Imp. B (EP), 1M-3PP
IUPAC Name N-methyl-3-phenylpropan-1-amine
SMILES CNCCCc1ccccc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H15N/c1-11-9-5-8-10-6-3-2-4-7-10/h2-4,6-7,11H,5,8-9H2,1H3
Formula Weight 149.1204
Chemical Name or Material methyl(3-phenylpropyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.