missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-4-(1H-pyrazol-1-ylmethyl)benzylamine, 97%, Thermo Scientific™

Product Code. 10245884
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10245884 250 mg Amber Glass Bottle 250mg
10787095 1 g Amber Glass Bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10245884 Supplier Thermo Scientific Maybridge Supplier No. CC56046CB
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 892502-08-8
Molecular Formula C12H15N3
Molecular Weight (g/mol) 201.27
MDL Number MFCD08690302
InChI Key BQLXMVISWVMAID-UHFFFAOYSA-N
Synonym n-methyl-4-1h-pyrazol-1-ylmethyl benzylamine, methyl 4-pyrazol-1-ylmethyl phenyl methyl amine, methyl 4-1h-pyrazole-1-yl methylbenzyl amine, methyl 4-pyrazolylmethyl phenyl methyl amine, benzenemethanamine,n-methyl-4-1h-pyrazol-1-ylmethyl, n-methyl-1-4-1h-pyrazol-1-ylmethyl phenyl methanamine, 1-4-1h-pyrazol-1-yl methyl phenyl-n-methylmethanamine, n-methyl-1-4-1h-pyrazol-1-yl methyl phenyl methanamine, 3vr
PubChem CID 18525877
IUPAC Name N-methyl-1-[4-(pyrazol-1-ylmethyl)phenyl]methanamine
SMILES CNCC1=CC=C(CN2C=CC=N2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 201.27
Percent Purity 97%
Chemical Name or Material N-methyl-4-(1H-pyrazol-1-ylmethyl)benzylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.