missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl Metribuzin, TRC

Product Code. 30469008
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30469008 2.5 g 2.5g
30470266 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30469008 Supplier Toronto Research Chemicals Supplier No. TRCM3202952.5G

Please to purchase this item. Need a web account? Register with us today!

This item requires a price update prior to being ordered.

Request a Price for this item here.
, 02 950 59 478

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56742-45-1
Molecular Formula C9 H16 N4 O S
Molecular Weight (g/mol) 228.31
Synonym 6-(1,1-Dimethylethyl)-4-(methylamino)-3-(methylthio)-1,2,4-triazin-5(4H)-one, 6-tert-Butyl-4-(methylamino)-3-(methylthio)-1,2,4-triazin-5(4H)-one
IUPAC Name 6-tert-butyl-4-(methylamino)-3-methylsulfanyl-1,2,4-triazin-5-one
SMILES CNN1C(=NN=C(C1=O)C(C)(C)C)SC

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C9H16N4OS/c1-9(2,3)6-7(14)13(10-4)8(15-5)12-11-6/h10H,1-5H3
Formula Weight 228.1045
Chemical Name or Material Metribuzin-N-methyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.