missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-(2-thien-2-ylbenzyl)amine, 97%, Thermo Scientific™

Product Code. 10767875
Change view
Click to view available options
Quantity:
250 mg
5 g
Unit Size:
250mg
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10767875 5 g 5g
10167552 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10767875 Supplier Thermo Scientific Maybridge Supplier No. CC35546DE
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 852180-66-6
Molecular Formula C12H13NS
Molecular Weight (g/mol) 203.303
MDL Number MFCD07368538
InChI Key MRKJJEJYTBOUTH-UHFFFAOYSA-N
Synonym n-methyl-n-2-thien-2-ylbenzyl amine, n-methyl-1-2-thiophen-2-ylphenyl methanamine, n-methyl-2-thien-2-ylbenzylamine, methyl 2-thiophen-2-yl phenyl methyl amine, n-methyl-1-2-thiophen-2-yl phenyl methanamine, d0a2yg, n-methyl-n-2-thiophen-2-ylbenzyl amine, methyl 2-2-thienyl phenyl methyl amine, benzenemethanamine,n-methyl-2-2-thienyl, benzenemethanamine, n-methyl-2-2-thienyl
PubChem CID 7060565
IUPAC Name N-methyl-1-(2-thiophen-2-ylphenyl)methanamine
SMILES CNCC1=CC=CC=C1C2=CC=CS2

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 95%
Quantity 5 g
Formula Weight 203.31
Percent Purity 95%
Chemical Name or Material N-methyl-N-(2-thien-2-ylbenzyl)amine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.